Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

218683

Sigma-Aldrich

Casein-α, His•Tag® Fusion

Synonym(s):

Casein-α, Human, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice

storage temp.

−70°C

General description

Full-length, recombinant human casein-α fused to protein expressed with N-terminal His•Tag and S•Tag sequences at the N-terminus. It is qualified for use as a substrate for protein tyrosine kinases in in vitro assays and may also serve as a good substrate for proteases. It is expected to be phosphorylated at Ser71 by PKA, Ser89 by CK-2, and Tyr109 by EGFR kinase.
Full-length, recombinant human casein-α fused to protein expressed with N-terminal His•Tag and S•Tag sequences at the N-terminus. Qualified for use as a substrate for protein tyrosine kinases in in vitro assays and may also serve as a good substrate for proteases. It is expected to be phosphorylated at Ser71 by PKA, Ser89 by CK-2, and Tyr109 by EGFR kinase.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Irritant (B)

Physical form

In PBS, 4 M Urea, Protease Inhibitor Cocktail Set VII (Cat. No. 539138).

Other Notes

Dogru, M., et al. 2006 Prep. Biochem. Biotechnol.36, 165.
Kasuga, M., et al. 1983. J. Biol. Chem.258, 10973.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

218683-50UG:
2186838093:
218683-500UG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service