Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

204889-M

Millipore

Anti-Complement 5a Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

should not react with

rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Complement 5a, rabbit polyclonal, is monospecific for human C5a complement component. It is validated for ELISA, Western blotting, and ouchterlony.
Monospecific for human C5a complement component as determined by OT.
Rabbit polyclonal antibody supplied as undiluted serum. Recognizes complement component C5a.

Immunogen

Human

Application

ELISA (see comments)

Immunoblotting (see comments)

Ouchterlony (use undiluted)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Monospecific for complement component C5a as determined by ouchterlony. This antibody is also reported to work for ELISA and immunoblotting. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

204889-1ML-M:
204889-ML:
204889-1ML:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service