Skip to Content
Merck
All Photos(1)

Documents

Safety Information

204883

Sigma-Aldrich

Complement C1-Esterase Inhibitor, Human

Complement C1-Esterase Inhibitor, Human, is a native, single-chain glycoprotein that is involved in regulation of complement, coagulation, fibrinolytic and contact systems of circulating plasma.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, albumin, C3, C4, or ceruloplasmin, trace

shipped in

wet ice

storage temp.

−70°C

General description

Native, human C1-esterase. Complent C1-esterase inhibitor purified from human plasma. Single chain glycoprotein present in normal human serum at a concentration of 180 µg/ml. Involved in regulation of complement, coagulation, fibrinolytic and contact (kinin-forming) systems of circulating blood plasma. It is the only known inhibitor of activated C1r and C1s complement enzymes. Persons with either hereditary or acquired deficiencies of C1 esterase inhibitor are susceptible to recurrent attacks of angioedema.
Native, human C1-esterase. Single-chain glycoprotein present in normal human serum at 180 µg/ml. Involved in regulation of complement, coagulation, fibrinolytic and contact (kinin-forming) systems of circulating blood plasma. The only known inhibitor of activated C1r and C1s complement enzymes. The presence of either hereditary or acquired deficiencies of C1 inhibitor produce susceptibility to recurrent attacks of angioedema.

Biochem/physiol Actions

≥80% of C1-esterase inhibitor will bind activated C1s.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.4, sterile-filtered through a 0.22 µm filter.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Bowen, B., et al. 2001. Clin. Immunol.98, 157.
Davis, A.E. 1988. Ann. Rev. Immunol. 6, 595.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

204883-1MG:
204883-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service