Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

203303

Sigma-Aldrich

Bisindolylmaleimide V

A cell-permeable negative control compound for protein kinase C inhibition studies (IC₅₀ >100 µM).

Synonym(s):

Bisindolylmaleimide V, 2,3- bis(1H-Indol-3-yl)-N-methylmaleimide, Ro 31-6045, 2,3-bis(1H-Indol-3-yl)-N-methylmaleimide, Ro 31-6045

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15N3O2
CAS Number:
Molecular Weight:
341.36
UNSPSC Code:
12352200
Pricing and Inventory is not available. Please check the Merck website for more information.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

red-orange

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C21H15N3O2/c1-24-20(25)18(14-10-22-16-8-4-2-6-12(14)16)19(21(24)26)15-11-23-17-9-5-3-7-13(15)17/h2-11,22-23H,1H3

InChI key

SWAWYMIKGOHZMR-UHFFFAOYSA-N

General description

A cell-permeable negative control compound for protein kinase C inhibition studies (IC50 >100 µM). Blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6kin vivo (IC50 8 µM).
A cell-permeable negative control compound for protein kinase C inhibition studies (IC50 >100 µM). Blocks the activation of p70s6k/p85s6kin vivo (IC50 = 8 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Negative control compound for protein kinase C inhibition
Product does not compete with ATP.
Reversible: no
Target IC50: >100 µM as negative control compound for protein kinase C inhibition; 8 µM in blocking the activation of mitogen-stimulated protein kinase p70s6k/p85s6k in vivo

Warning

Toxicity: Standard Handling (A)

Other Notes

Marmy-Conus, N., et al. 2002. FEBS Lett. 26094 (In Press) 1-6.
Davis, P.D., et al. 1992. J. Med. Chem.35, 177.
Toullec, D., et al. 1991. J. Biol. Chem.266, 15771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

203303-MG:
203303-1.1ML:
203303-1MG:
203303-250UG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen R Reeves et al.
Experimental physiology, 92(6), 1057-1066 (2007-08-07)
Protein kinase C (PKC) is a broadly expressed and critically important signalling protein with a wide range of functional roles, including central components of respiratory control. For example, systemic and targeted administration of PKC inhibitors within the nucleus of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service