Skip to Content
Merck
All Photos(1)

Documents

Safety Information

20-132

Sigma-Aldrich

PKA/CaMK Inhibitor Cocktail

The PKA/CaMK Inhibitor Cocktail controls the biological activity of PKA/CaMK. This small molecule/inhibitor is primarily used for Biochemicals applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Application

For use in kinase assays.

Biochem/physiol Actions

Inhibitor Type: Kinase
Protein Target: PKA C-alpha

Physical form

20mM MOPS, pH 7.2, 25mM β-glycerophosphate, 1mM sodium orthovanadate, 1mM dithiothreitol, 1mM CaCl2

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

20-132:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ubiquitin-proteasome pathway and enhanced activity of NF-kappaB in gastric carcinoma.
Lingfei Wu,Zejin Pu,Jialin Feng,Guoping Li,Zhichao Zheng,Wenlv Shen
Journal of Surgical Oncology null
Redox control of 20S proteasome.
Bertrand Friguet,Anne-Laure Bulteau,Mariangela Conconi,Isabelle Petropoulos
Methods in Enzymology null
Huiqing Zeng et al.
PloS one, 5(12), e15900-e15900 (2011-01-07)
Intracellular transduction of Hedgehog (Hh) signals in mammals requires functional primary cilia. The Hh signaling effectors, the Gli family of transcription factors, and their negative regulator, Suppressor of Fused (Sufu), accumulate at the tips of cilia; however, the molecular mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service