Skip to Content
Merck
All Photos(1)

Documents

Safety Information

1.93270

Supelco

Sodium fluoride

for analysis EMPARTA® ACS

Synonym(s):

Sodium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
12352302

grade

ACS reagent

vapor pressure

1.4 mmHg ( 0 °C)

mp

993 °C (lit.)

storage temp.

room temp

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93270.0521: 500 g, Minimum order quantity: 20

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

1932700521:
IN2RCMS193270:
1932700500:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service