Skip to Content
Merck
All Photos(4)

Key Documents

Safety Information

17-10082

Sigma-Aldrich

PureEpi Chromatin Preparation and Optimization Kit

The PureEpi Chromatin Preparation & Optimization kit provides a complete set of reagents allowing the preparation of multiple chromatin samples from starting materials such as cultured cells or tissues.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.52

Quality Level

manufacturer/tradename

PureEpi
Upstate®

application(s)

sample preparation

shipped in

dry ice

General description

The PureEpi Chromatin Preparation and Optimization kit provides a complete set of reagents allowing the preparation of multiple chromatin samples from starting materials such as cultured cells or tissues. A detailed protocol is provided that includes examples of the many variables involved in making chromatin on multiple instruments and how they may affect ChIP results.

Features & Benefits
  • Complete set of materials for up to 12 chromatin preparations
  • Optimized streamlined protocol with only a single buffer for sonication, IP, or wash
  • Protocols for using cells or tissues
  • Compatible with Magna ChIP HiSens and EZ-Magna ChIP HiSens chromatin immunoprecipitation kits.
  • Compatible with ChIPAb+ validated antibody and primer sets

Application

Research Category
Epigenetics & Nuclear Function
The PureEpi Chromatin Preparation & Optimization kit provides a complete set of reagents allowing the preparation of multiple chromatin samples from starting materials such as cultured cells or tissues.

Packaging

Kit capacity: 12 chromatin preparations

Components

10X Glycine

Nuclei Isolation Buffer

SCW Buffer (Sonication/ChIP/Wash)

Crosslink Reversal Buffer

Protease Inhibitor Cocktail II

Physical form

Two boxes containing all necessary reagents to perform 12 chromatin preparations using 15 cm plates of cultured cells or tissue samples.

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 1 year from date of shipment when stored as directed.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service