Skip to Content
Merck
All Photos(2)

Documents

Safety Information

16-263

Sigma-Aldrich

Anti-Myc Tag Antibody, clone 9E10, Alexa Fluor 555 conjugate

clone 9E10, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Related Categories

Specificity

Myc and recombinant proteins containing the Myc epitope tag.

Immunogen

Synthetic peptide corresponding to amino acids 408-437 of the Leucine zipper region of human Myc (AEEQKLISEEDLLRKRREQLKHKLEQLRNS).

Application

Detect Myc Tag using this Anti-Myc Tag Antibody, clone 9E10, Alexa Fluor 555 conjugate validated for use in WB & IC.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Routinely evaluated by immunocytochemistry on HeLa cells expressing myc-tagged protein.

Physical form

Protein G Purified

Storage and Stability

1 year at 4°C from date of shipment. Do not store diluted material. Do not freeze.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

16-263:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determinants of the HIV-1 core assembly pathway.
Lopez, CS; Eccles, JD; Still, A; Sloan, RE; Barklis, RL; Tsagli, SM; Barklis, E
Virology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service