Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

14-909-M

Sigma-Aldrich

MELK Protein, active, 0 0

Active, recombinant human MELK (amino acids 1 -340), containing an N-terminal GST tag, for use in Kinase Assays.

Synonym(s):

HPK38, PK38, pEg3 kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected insect cells

mol wt

Mw 66 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... MELK(9833)

General description

Maternal embryonic leucine zipper kinase (MELK) is a conserved serine/threonine kinase involved in a variety of cellular functions, including proliferation, differentiation and apoptosis. Its levels are elevated in a number of tumour types and it may be a prognosis marker for some cancers.
Product Source: Recombinant protein of the human MELK gene expressed by baculovirus in insect cells
Recombinant human MELK (amino acids 1 - 340), containing an N-terminal GST tag

Application

Research Category
Signaling

Biochem/physiol Actions

Target Sub-Family: CAMK

Packaging

Also available in 250μg size - call for pricing and availability and reference catalog number 14-909M when ordering the 250μg size

Quality

Routinely evaluated by phosphorylation of KKLNRTLSFAEPG

Physical form

Purified using glutathione-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

14-909:
14-909M:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service