Skip to Content
Merck
All Photos(1)

Documents

Safety Information

14-746M-M

Sigma-Aldrich

Src (1-530) Protein, active, 250 µg

Active, N-terminal 6His-tagged, recombinant, human Src, amino acids 1-530, expressed by baculovirus in Sf21 insect cells. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 61.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

GenBank accession no.

Gene Information

human ... SRC(6714)

General description

N-terminal 6His-tagged, recombinant, human Src, amino acids 1-530, expressed by baculovirus in Sf21 insect cells.
Product Source: Expressed by baculovirus in Sf21 insect cells.
The mutant form of Src (Src, 1-530) has been identified in cases of advanced colon cancer. This truncating mutation generates an open active form of Src that has been found to be transforming, tumorigenic and metastasis promoting.

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: Src
Target Sub-Family: TK

Packaging

Also available in 10 μg size (cat#14-746) and in bulk (cat# 14-746-K).

Physical form

50mM Tris-HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution
Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C from date of shipment.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

14-746M:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service