Skip to Content
Merck
All Photos(1)

Documents

14-511-M

Sigma-Aldrich

Aurora A Protein, active, 10 µg

Active, N-terminal His6-tagged, recombinant full-length human Aurora A, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 46.9 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

General description

N-terminal His6-tagged, recombinant full-length human Aurora A
Product Source: expressed in Sf21 cells

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: Aurora-A
Target Sub-Family: AGC

Packaging

Also available in 250μg size as catalog # 14-511M - Call for pricing and availability.

Quality

routinely evaluated by phosphorylation of Kemptide

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service