Skip to Content
Merck
All Photos(1)

Documents

144216

Universal Battery Charger

for use with RCS® Isolator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® Isolator

shipped in

ambient

storage temp.

room temp

General description

940820 (RCS Isolator)
For charging the integrated 12 V NiMH batteries of the RCS Isolator operating unit

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service