Skip to Content
Merck
All Photos(2)

Documents

Safety Information

1.40118

Supelco

Extran® AP 18

greener alternative

liquid, mild alkaline concentrate

Synonym(s):

TM=["EXTRAN"] AP 18 cleaning agent; TM=["EXTRAN"] AP 18 detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161902
NACRES:
NA.21

Quality Level

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

13.5 (20 °C in H2O)

greener alternative category

storage temp.

15-25°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Application

Extran® AP 18 is a mild alkaline laboratory cleaning agent used for gentle cleaning of alkali-sensitive items. It does not foam even during heavy agitation in a washing machine. Extran® AP 18 is a liquid main cleaning agent with mild alkaline properties for automatic dosage.
Its properties includes:
  • surfactant-free
  • chlorine-free
  • phosphate-free
  • free from odorants/dyestuffs

Linkage

Replaces: EX1009; 1.40001; 1.40001.2500; 1.40001.5000; 1.40001.9010

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service