Skip to Content
Merck
All Photos(1)

Documents

Safety Information

1.16981

Supelco

Ascorbic Acid Test Strips, reflectometric

25-450 mg/L (ascorbic acid), REFLECTOQUANT®

Synonym(s):

Ascorbic Acid chemical test strips, Ascorbic Acid test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Ascorbic Acid Test, reflectometric, 25-450 mg/L (ascorbic acid), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

ascorbic acid

measuring range

25-450 mg/L (ascorbic acid)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Ascorbic Acid (vitamin C) counts as an essential quality characteristic in many foods. The content of ascorbic acid is measured during the production and filling procedures in the fruit- and vegetable-processing sector as a measure to guarantee compliance of the minimum required values for preservation. The Reflectoquant Ascorbic acid test strips are suitable for quickly measuring levels of natural ascorbic acid. Ascorbic acid reduces yellow molybdophosphoric acid to phosphormolybdenum blue that is deter mined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the ascorbic acid concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for ascorbic acid.
Method: reflectometric with test strips 25 - 450 mg/l Reflectoquant®

Linkage

Replaces: 16981-1; 16981

Analysis Note

function control: conforms
Assay
- standard 1:
45 - 55 mg/l
- standard 2: 180 - 220 mg/l
- standard 3: 360 - 440 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Notified Names

Substances Subject to be Notified Names


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service