Skip to Content
Merck
All Photos(1)

Documents

Safety Information

116850

Sigma-Aldrich

Adenylyl Cyclase Type V Inhibitor, NKY80

The Adenylyl Cyclase Type V Inhibitor, NKY80, also referenced under CAS 299442-43-6, controls the biological activity of Adenylyl Cyclase Type V. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Adenylyl Cyclase Type V Inhibitor, NKY80, 2-Amino-7-(furanyl)-7,8-dihydro-5(6H)-quinazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N3O2
CAS Number:
Molecular Weight:
229.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale orange

solubility

DMSO: 200 mg/mL
methanol: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C12H11N3O2/c13-12-14-6-8-9(15-12)4-7(5-10(8)16)11-2-1-3-17-11/h1-3,6-7H,4-5H2,(H2,13,14,15)

InChI key

SOJUSNIBPPMLCC-UHFFFAOYSA-N

General description

A cell-permeable quinazolinone containing a non-nucleoside compound that acts as a potent, selective inhibitor of adenylyl cyclase (AC) type V isoform (IC50 = 8.3 µM, 132 µM and 1.7 mM for type V, III and II, respectively) in the presence of Gsα GTPγS-Forskolin. Displays ~210-fold greater selectivity for type V over the type II isoform. The inhibition is non-competitive with respect to ATP and Forskolin. Regulates the AC catalytic activity in heart and lung tissues.
A cell-permeable, non-nucleoside compound that acts as a potent, and selective inhibitor of adenylyl cyclase (AC) type V (IC50 = 8.3 µM, 132 µM and 1.7 mM for type V, III and II, respectively) in the presence of G5α GTPγS-Forskolin. Displays ~210-fold greater selectivity for type V over the type II isoform. The inhibition is non-competitive with respect to ATP and Forskolin. Reported to regulate AC catalytic activity in heart and lung tissues.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Adenylate cyclase (AC) type V isoform
Product does not compete with ATP.
Reversible: no
Target IC50: 8.3 µM, 132 µM and 1.7 mM for type V, III and II, of adenylyl cyclase (AC), respectively, in the presence of Gsα GTPγS-Forskolin

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Onda, T., et al. 2001. J. Biol. Chem.276, 47785.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

116850-5MG:
116850-1.1ML:
116850-5UNMG:
116850-25MG:
116850-MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katja Baur et al.
Life science alliance, 7(7) (2024-05-09)
We recently reported that growth/differentiation factor 15 (GDF15) and its receptor GDNF family receptor alpha-like (GFRAL) are expressed in the periventricular germinal epithelium thereby regulating apical progenitor proliferation. However, the mechanisms are unknown. We now found GFRAL in primary cilia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service