Skip to Content
Merck
All Photos(1)

Key Documents

1.11926

Millipore

Yeast extract

for biotechnology Fermtech®

Synonym(s):

Yeast extract, Extract of yeast

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
232-387-9
Pricing and Inventory is not available. Please check the Merck website for more information.

biological source

yeast

Quality Level

form

solid

autoignition temp.

>200 °C

pH

7.0 (20 °C, 10 g/L in H2O)

solubility

410 g/L

density

0.5 g/cm3 at 20 °C

bulk density

490 kg/m3

application(s)

microbiology

storage temp.

15-25°C

Analysis Note

Appearance: Brownishyellow fine powder
Identity: passes test
Total nitrogen (N) (calculated on dried substance): ≥ 8.4 %
Amino nitrogen: ≥ 4.0 %
pH-value: 5.5 - 6.0
Loss on drying: ≤ 6.0 %
Ash (without salt): ≤ 13.0 %

Legal Information

FERMTECH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service