Skip to Content
Merck
All Photos(1)

Key Documents

1.09795

Supelco

n-Undecane

for analysis EMSURE®

Synonym(s):

n-Undecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
MDL number:
UNSPSC Code:
12191502
EC Index Number:
214-300-6
Pricing and Inventory is not available. Please check the Merck website for more information.

vapor pressure

0.6 hPa ( 20 °C)

Quality Level

product line

EMSURE®

Assay

≥99% (GC)

form

liquid

autoignition temp.

220 °C

expl. lim.

0.6-6.5 % (v/v)

impurities

≤0.00008 meq/g Acidity
≤0.005% Sulfated ash
≤0.01% Water

kinematic viscosity

1.6 cSt(20 °C)

mp

-26 °C

transition temp

flash point 60 °C

density

0.74 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C11H24/c1-3-5-7-9-11-10-8-6-4-2/h3-11H2,1-2H3

InChI key

RSJKGSCJYJTIGS-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

60 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service