Skip to Content
Merck
All Photos(1)

Documents

1.03865

Millipore

GranuCult® prime OF (Oxidative/Fermentative) Base

according to ISO (21528)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

according to ISO (21528)

Quality Level

form

powder

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

antibiotic activity spectrum

Gram-negative bacteria

application(s)

food and beverages
pharmaceutical

suitability

Alcaligenes spp.
Chromobacterium spp.
Moraxella spp.
Pasteurella spp.
Acinetobacter spp.
Aeromonas spp.
Bordetella spp.
Campylobacter spp.
Enterobacter spp.
Escherichia coli
Pseudomonas spp.
Salmonella spp.
Shigella spp.
Vibrio spp.
Yersinia spp.
flavobacteria
nonselective for bacteria

General description

The GranuCult® prime OF (Oxidative/Fermentative) medium is designed for the confirmation and identification of gram-negative bacteria based on oxidative or fermentative metabolism of carbohydrates. The medium is supplemented with glucose for the confirmation of Enterobacteriaceae present in the test samples. The pH indicator, bromothymol blue indicates the degradation of the carbohydrate by a color change reaction (blue to yellow). The reaction can take place either aerobically or anaerobically.

Application

The GranuCult® prime OF (Oxidative/Fermentative) medium is used to grow bacteria which can metabolize carbohydrates. The medium can also promote growth of all aerobic, anerobic and facultative aerobic bacteria.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service