Skip to Content
Merck
All Photos(3)

Documents

Safety Information

07-329-AF488

Sigma-Aldrich

Anti-acetyl-Histone H4 (Lys16) Antibody, Alexa Fluor 488

from rabbit, ALEXA FLUOR 488

Synonym(s):

Histone H4, acetyl-Histone H4 (Lys16), AlexaFluor 488

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys16)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

General description

Histone H4 (UniProt P02309) is encoded by the HHF1 (ORF YBR0122; Gene ID 852294) and HHF2 (ORF N2752; Gene ID 855701) genes in yeast species. Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specificity

Broad species cross-reactivity is expected.
H4K16ac specificity of the unconjugated antibody (Cat. No. 07-329) has been confirmed using the Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and the Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).

Immunogen

Epitope: H4K16ac
Ovalbumin-conjugated, synthetic peptide (…GAAcKRH …) in which AcK corresponds to residue 16 of yeast histone H4.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected acetyl-Histone H4 (Lys16) in A431 and NIH/3T3 cells.
The unconjugated antibody (Cat. No. 07-329) is shown to be suitable also for ChIP, ChIP-seq, Western blotting, multiplex assay, dot blot applications.
This Anti-acetyl-Histone H4 (Lys16) Antibody, AlexaFluor 488 is validated for use in Immunocytochemistry for the detection of acetyl-Histone H4 (Lys16).

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected acetyl-Histone H4 (Lys16) in HeLa cells.

Target description

~10 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

07-329-AF488:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Pessoa Rodrigues et al.
Science advances, 6(21), eaaz4815-eaaz4815 (2020-07-17)
Self-renewal and differentiation of hematopoietic stem cells (HSCs) are orchestrated by the combinatorial action of transcription factors and epigenetic regulators. Here, we have explored the mechanism by which histone H4 lysine 16 acetyltransferase MOF regulates erythropoiesis. Single-cell RNA sequencing and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service