Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

07-2114

Sigma-Aldrich

Anti-YAP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Yorkie homolog, 65 kDa Yes-associated protein, YAP65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... YAP1(10413)

General description

YAP1 transcription factor is known to regulate the response to oxidative stress in S. cerevisiae. YAP1 is activated by increased levels of reactive oxygen species and moves quickly to the nucleus where it regulates the expression of up to 70 genes. Intramolecular disulphide bond formation is thought to be the mechanism of transcription factor control in YAP1. Tumor suppressor RASSF1A releases YAP1, which is then free to interact with p73 and transcription of pro-apoptotic puma begins. In the skin, YAP1 is the transcriptional effector of the Hippo signaling pathway and acts as the determinant of proliferative capacity of epidermal stem cells.

Immunogen

GST-tagged recombinant protein corresponding to human YAP1.

Application

Research Category
Signaling
Research Sub Category
Oxidative Stress
This YAP1 antibody is validated for use in WB for the detection of the YAP1 protein.

Quality

Evaluated by Western Blot in human liver tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected YAP1 on 10 µg of human liver tissue lysate.

Target description

~60 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

07-2114:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service