Skip to Content
Merck
All Photos(3)

Documents

Safety Information

06-1130-I

Sigma-Aldrich

Anti-RNF168 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

E3 ubiquitin-protein ligase RNF168, hRNF168, RING finger protein 168

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RNF168(165918)

General description

E3 ubiquitin-protein ligase RNF168 (UniProt Q8IYW5; also known as hRNF168, RING finger protein 168) is encoded by the RNF168 gene (Gene ID 165918) in human. Ubiquitylation/Ubiquitination is an essential early signal in the DNA damage repair process. Multiple E3 ligases, including RNF2 (RING2), RNF8 and RNF168, mediate the addition of K63-linked polyubiquitin chains to histone 2A (H2A) and H2AX, which in turn recruits DNA damage repair proteins, e.g. RAP80, BRCA1. In addition, SUMOylation and NEDDylation are also involved in the DNA damage response. RNF168 is an a dual specificity E3 ligase that mediates both the ubiquitination and NEDDylation of H2A and H2AX. The two types of modifications compete against each other and NEDD8 modification of H2A and H2AX blocks the recruitment of BRCA1 at DNA damage repair sites. RNF168 itself is also subjected to NEDD8 modification and NEDDylation of RNF168 is necessary for its ubiquitin ligase activity. Inhibition of RNF168 NEDDylation impairs its interaction with E2 enzyme Ubc13 (UBE2N). Likewise, downregulating RNF168 NEDDylation by the deNEDDylating enzyme NEDP1, or due to mutations of the NEDD8-conjugating enzyme UBC12, decreases H2A and H2AX ubiquitylation. RNF168 Mutations are linked to RIDDLE (radiosensitivity, immunodeficiency dysmorphic features and learning difficulties) syndrome.

Specificity

This polyclonal antibody detected nuclear RNF168 immunoreactivity in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized human B-cell hybridoma SA13.

Immunogen

GST-tagged full-length human RNF168.

Application

Anti-RNF168, Cat. No. 06-1130-I, is a highly specific rabbit polyclonal antibody that targets RNF168 and has been tested in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected nuclear RNF168 immunoreactivity in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized human B-cell hybridoma SA13.

Immunoprecipitation Analysis: 20 µL from a representative lot immunoprecipitated RNF168 from 500 µg of human B-cell hybridoma SA13 cell lysate.

Quality

Evaluated by Western Blotting in SA13 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RNF168 in 10 µg of human B-cell hybridoma SA13 cell lysate.

Target description

~75 kDa observed. 65.02 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 06-1130

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

06-1130-I:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John J Krais et al.
Nucleic acids research, 48(8), 4298-4308 (2020-03-18)
The RNF168 E3 ubiquitin ligase is activated in response to double stranded DNA breaks (DSBs) where it mono-ubiquitinates γH2AX (ub-H2AX). RNF168 protein expression and ubiquitin signaling are finely regulated during the sensing, repair and resolution of DNA damage in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service