Skip to Content
Merck
All Photos(2)

Documents

Safety Information

06-1116

Sigma-Aldrich

Anti-IL-33R ST2 (IL1RL1) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

interleukin 1 receptor-like 1, interleukin-1 receptor-like 1, growth stimulation-expressed, homolog of mouse growth stimulation-expressed, interleukin 1 receptor-related protein, Protein ST23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Interleukin-1 receptor-like 1 (IL-33R ST2) is a member of the interleukin-1 receptor family, and is comprised of three Ig-like C2-type domains and a single TIR domain. IL-33R ST2 functions as a receptor for IL-33, and, following phosphorylation with MAPKa/ERK2, MAPK3/ERK1, MAPK8 and/ore MAPK14, serves to recruit IRAK1, IRAk4, MYD88, and TRAF6. It is also thought to have a potential role in the function of helper T-cells. High expression has been reported in colon, kidney, lung, placenta, skeletal muscle, small intestines, and the stomach. Isoform A has been noted to be more prevalent in colon, lung, placenta, testis and stomach, while Isoform B has greater presence in brain, liver and kidney.

Specificity

This antibody recognizes the extracellular domain of IL-33R ST2.

Immunogen

Epitope: Extracellular Domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human IL-33R ST2.

Application

Anti-IL-33R ST2 (IL1RL1) Antibody is an antibody against IL-33R ST2 (IL1RL1) for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:300 dilution from a previous lot detected IL-33R ST2 in normal human thymus tissue.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blot in human kidney tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected IL-33R ST2 in 10 µg of human kidney tissue lysate.

Target description

~30 kDa observed.
UniProtKB/Swiss-Prot Q01638 (ILRL1_HUMAN) entry describes 3 isoforms produced by alternative splicing: Isoform A at 63.358 kDa, Isoform B at 36.993 kDa, Isoform C at 29.951 kDa The ~37 kDa product is the secreted form.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human kidney tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

06-1116:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service