Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

05-733

Sigma-Aldrich

Anti-SETA/CIN85/Ruk/SH3KBPI Antibody, clone 84

clone 84, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

84, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SH3KBP1(30011)

Related Categories

Specificity

SETA

Immunogen

6His and HA tagged fusion protein corresponding to full length rat SETA

Application

Detect SETA/CIN85/Ruk/SH3KBPI using this Anti-SETA/CIN85/Ruk/SH3KBPI Antibody, clone 84 validated for use in IP, WB & IC.

Quality

routinely evaluated by immunoblot on RIPA lysates from NG108-15 cells

Target description

85kDa

Physical form

Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

05-733:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CIN85 associates with endosomal membrane and binds phosphatidic acid.
Jing Zhang, Xiudan Zheng, Xiao Yang, Kan Liao, Jing Zhang, Xiudan Zheng, Xiao Yang, Kan Liao
Cell research null
Susan Finniss et al.
Biochemical and biophysical research communications, 325(1), 174-182 (2004-11-04)
SETA/CIN85/Ruk is a multifunctional adaptor protein involved in signal transduction and attenuation downstream of receptor tyrosine kinases. It has a modular structure, and various isoforms that combine different protein-protein interaction domains have been proposed based on cDNA analysis. As a
Tom Büchse et al.
Molecular & cellular proteomics : MCP, 10(10), M110-M110 (2011-07-05)
The Cbl-interacting 85-kDa protein (CIN85) plays an important role as a negative regulator of signaling pathways induced by receptor tyrosine kinases. By assembling multiprotein complexes this versatile adaptor enhances receptor tyrosine kinase-activated clathrin-mediated endocytosis and reduces phosphatidylinositol-3-kinase-induced phosphatidylinositol-3,4,5-trisphosphate production. Here
B Chen et al.
The Journal of biological chemistry, 275(25), 19275-19281 (2000-06-20)
Expression of the src homology 3 (SH3) domain-containing expressed in tumorigenic astrocytes (SETA) gene is associated with the tumorigenic state in astrocytes. SETA encodes a variety of adapter proteins containing either one or two SH3 domains, as suggested by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service