Skip to Content
Merck
All Photos(1)

Documents

Safety Information

04-1556

Sigma-Aldrich

Anti-MDM4 Antibody, clone 7A8

clone 7A8, from mouse

Synonym(s):

Double minute 4 protein, MDM4-related protein 1, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding

protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding pr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A8, monoclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDM4(4194)
mouse ... Mdm4(17248)

General description

MDM4 (sometimes referred to as MDMX) is a member of the MDM family proteins. It is known to inhibit p53 and p73 mediated cell cycle arrest and apoptosis. MDM4 is amplified or overexpressed in 10-20% of tumors and 65% of retinoblastomas. The full-length protein has significant homology to MDM2 and can also bind to and inhibit p53. In both MDM2 and MDM4, this interaction with p53 occurs via a binding domain located in the N-terminal region of the protein. Other similar domains include the C-terminal RING finger motif. Although these proteins share similar domains, MDM2 and MDM4 are non-redundant p53 inhibitors, since normal levels of either protein does not compensate for the loss of the other. Recent evidence suggests complementary activities for these proteins with MDM4 regulating p53 activity, and MDM2 playing a key role in p53 stability.

Specificity

This antibody recognizes MDM4.

Immunogen

Epitope: Unknown
His-tagged recombinant protein corresponding to mouse MDM4.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Transcription Factors

Apoptosis - Additional
Use Anti-MDM4 Antibody, clone 7A8 (mouse monoclonal antibody) validated in WB to detect MDM4 also known as Double minute 4 protein, MDM4-related protein 1, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse).

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected MDM4 in 10 µg of HeLa cell lysate.

Target description

~ 80/27 kDa Observed. There are multiple forms for MDM4. Up to 7 different variants have been reported. Full legth of MDM4 is ~80kDa The short form of MDM4 (MDM4-S) migrates at approximately 17 and 27kDa (R. Rallapalli, G. Strachan, B. Cho, W. Mercer‡, and DJ. Hall, 1999, A Novel MDMX Transcript, THE JOURNAL OF BIOLOGICAL CHEMISTRY, 274(12), 8299–8308)

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

04-1556:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiahao Shi et al.
Oncogenesis, 11(1), 37-37 (2022-07-03)
p53 is the most highly mutated tumor suppressor across multiple types of human cancers. The level and function of p53 are fine-tuned through multifaced mechanisms in which the protein-protein interaction between p53 and MDM2 is considered as a major circuit.
Chiara Naro et al.
Journal of experimental & clinical cancer research : CR, 43(1), 58-58 (2024-02-28)
Advanced prostate cancer (PC) is characterized by insensitivity to androgen deprivation therapy and chemotherapy, resulting in poor outcome for most patients. Thus, advanced PC urgently needs novel therapeutic strategies. Mounting evidence points to splicing dysregulation as a hallmark of advanced
Olan Jackson-Weaver et al.
Cell reports, 31(10), 107739-107739 (2020-06-11)
Epicardial cells are cardiac progenitors that give rise to the majority of cardiac fibroblasts, coronary smooth muscle cells, and pericytes during development. An integral phase of epicardial fate transition is epithelial-to-mesenchymal transition (EMT) that confers motility. We uncover an essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service