03-0199-00
SMC® Human NPTX2 High Sensitivity Immunoassay Kit
Synonym(s):
NPTX2 high sensitive immunoassay, NPTX2 single molecule counting kit,, Neuronal pentraxin 2 immuno assay kit
About This Item
Recommended Products
manufacturer/tradename
SMC®
assay range
sensitivity: 0.58 pg/mL
(Lower Limit of Quantification)
inter-assay cv: 5%
intra-assay cv: 6%
(five normal plasma samples run in triplicate by 3 different operators on 3 different days)
technique(s)
single molecule counting: suitable
Related Categories
General description
Specificity
Specificity to the following species samples were tested with the follow results:
Non human primate – 100%
Mouse – 100%Rat – 100%
Canine – 100%
Feline – 100%
Cross ReactivityN/A
Accuracy
Minimum of 5 serum samples spiked with average recovery of 99% with a range of 93-103%
Minimum of 5 plasma samples spiked with average recovery of 104% with a range of 97-118%
Application
Packaging
Components
- Assay Buffer
- NPTX2 Coated Beads
- Standard Diluent
- NPTX2 Detection Antibody
- NPTX2 Standard
- 10X Wash Buffer
- Buffer D
- Elution Buffer B
Other Notes
- Improved options for sample prep and incubation
- Lyophilization protein standard for uniform shipping and storage condition
- Increased reagent volumes to accommodate automation
- Able to achieve assay specifications on both SMCxPRO® and Erenna
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
03-0199-00:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Neuroscience biomarker research expands with high-sensitivity immunoassays for low-abundant biomarkers.
Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service