Skip to Content
Merck
All Photos(1)

Key Documents

MAN326367645

Manchester Organics

2,4,5-Trichlorophenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
225.27
MDL number:
Pricing and availability is not currently available.

Iupac Name

2,4,5-trichlorophenylboronic acid

Inchi Code

1S/C6H4BCl3O2/c8-4-2-6(10)5(9)1-3(4)7(11)12/h1-2,11-12H

InChI key

FTLYMKDSHNWQKD-UHFFFAOYSA-N

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/K22387

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library