MAN238930187
Manchester Organics
7-Chloro-1,2,3,4-tetrahydroisoquinoline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
7-chloro-1,2,3,4-tetrahydroisoquinoline
Inchi Code
1S/C9H10ClN/c10-9-2-1-7-3-4-11-6-8(7)5-9/h1-2,5,11H,3-4,6H2
InChI key
BLRKQLICTKRDDZ-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Product Link
https://www.manchesterorganics.com/Z20057
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.