Skip to Content
Merck
All Photos(1)

Key Documents

KEY267019866

Key Organics / BIONET

ethyl 1-thia-4-azaspiro[4.5]decane-6-carboxylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
265.8
MDL number:

mp

183 - 185

Iupac Name

ethyl 1-thia-4-azaspiro[4.5]decane-6-carboxylate hydrochloride

Inchi Code

1S/C11H19NO2S.ClH/c1-2-14-10(13)9-5-3-4-6-11(9)12-7-8-15-11;/h9,12H,2-8H2,1H3;1H

InChI key

QRBRXLYGCLXWSE-UHFFFAOYSA-N

storage temp.

Store under inert gas

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/AE-0733

Melting Point

183 - 185

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/AE-0733


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library