Skip to Content
Merck
All Photos(1)

Key Documents

ENA638598597

Enamine

tert-butyl N-[(1-benzyl-1,2,3,4-tetrahydroquinolin-3-yl)methyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
352.48
MDL number:

Iupac Name

tert-butyl ((1-benzyl-1,2,3,4-tetrahydroquinolin-3-yl)methyl)carbamate

Inchi Code

1S/C22H28N2O2/c1-22(2,3)26-21(25)23-14-18-13-19-11-7-8-12-20(19)24(16-18)15-17-9-5-4-6-10-17/h4-12,18H,13-16H2,1-3H3,(H,23,25)

InChI key

JMCYGGKINFUVMD-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-345474

MSDS

https://www.enaminestore.com/catalog/EN300-345474


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library