Skip to Content
Merck
All Photos(1)

Key Documents

ENA444725547

Enamine

8-aza-9-boratricyclo[8.4.0.0,2,7]tetradeca-1(14),2,4,6,10,12-hexaen-9-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
195.03
MDL number:
Pricing and availability is not currently available.

mp

169-170

Iupac Name

dibenzo[c,e][1,2]azaborinin-6(5H)-ol

Inchi Code

1S/C12H10BNO/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14-13/h1-8,14-15H

InChI key

FXBGYQGNFKGWAU-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-203150

Melting Point

169-170

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-203150


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library