Skip to Content
Merck
All Photos(1)

Key Documents

ENA375227318

Enamine

ethyl 2-cyano-2-methyl-3-(pyridin-4-yl)propanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
218.26
MDL number:

Iupac Name

ethyl 2-cyano-2-methyl-3-(4-pyridinyl)propanoate

Inchi Code

1S/C12H14N2O2/c1-3-16-11(15)12(2,9-13)8-10-4-6-14-7-5-10/h4-7H,3,8H2,1-2H3

InChI key

XMLURGUDYJVFPJ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-111560

MSDS

https://www.enaminestore.com/catalog/EN300-111560


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library