ENA341294830
Enamine
2-methylmorpholine-4-sulfonyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-methyl-4-morpholinesulfonyl chloride
Inchi Code
1S/C5H10ClNO3S/c1-5-4-7(2-3-10-5)11(6,8)9/h5H,2-4H2,1H3
InChI key
FYKOGVZTHAJLMB-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-90511
MSDS
https://www.enaminestore.com/catalog/EN300-90511
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.