Skip to Content
Merck
All Photos(1)

Key Documents

ENA321763740

Enamine

1,3-diethyl 2-methanesulfonamidopropanedioate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
253.28
MDL number:
Pricing and availability is not currently available.

mp

74-75

Iupac Name

diethyl 2-[(methylsulfonyl)amino]malonate

Inchi Code

1S/C8H15NO6S/c1-4-14-7(10)6(8(11)15-5-2)9-16(3,12)13/h6,9H,4-5H2,1-3H3

InChI key

HSSFRIVDLKBXLS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-75600

Melting Point

74-75

MSDS

https://www.enaminestore.com/catalog/EN300-75600


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library