ENA152121134
Enamine
(1,2,3,4-tetrahydronaphthalen-1-yl)methanamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1,2,3,4-tetrahydro-1-naphthalenylmethylamine
Inchi Code
1S/C11H15N/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-2,4,7,10H,3,5-6,8,12H2
InChI key
FSEUPTSLGGJFNX-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-30476
Flash Point
124
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-30476
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.