Skip to Content
Merck
All Photos(1)

Key Documents

COMH93D66DF5

Combi-Blocks, Inc.

1-(3,5-Dibromopyridin-2-yl)thiourea

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
311
MDL number:

Iupac Name

1-(3,5-dibromopyridin-2-yl)thiourea

Inchi Code

1S/C6H5Br2N3S/c7-3-1-4(8)5(10-2-3)11-6(9)12/h1-2H,(H3,9,10,11,12)

InChI key

XIMYCSKDDBMLSY-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QG-2075

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QG-2075


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library