Skip to Content
Merck
All Photos(1)

Key Documents

COM964466629

Combi-Blocks, Inc.

Methyl 10-O-tert-Butyldimethylsilyl-oleanolate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
585
MDL number:

Iupac Name

methyl (4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-((tert-butyldimethylsilyl)oxy)-2,2,6a,6b,9,9,12a-heptamethyl-1,3,4,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-4a(2H)-carboxylate

Inchi Code

1S/C37H64O3Si/c1-31(2,3)41(12,13)40-29-17-18-34(8)27(33(29,6)7)16-19-36(10)28(34)15-14-25-26-24-32(4,5)20-22-37(26,30(38)39-11)23-21-35(25,36)9/h14,26-29H,15-24H2,1-13H3/t26-,27-,28+,29-,34-,35+,36+,37-/m0/s1

InChI key

PFXQUFDRMQZVJI-RRLKWQEHSA-N

storage temp.

Refrigerated

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?SH-6418

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?SH-6418


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library