Skip to Content
Merck
All Photos(1)

Key Documents

COM517298104

Combi-Blocks, Inc.

3-Bromo-5-phenylphenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
249.11
MDL number:

Iupac Name

5-bromo[1,1'-biphenyl]-3-ol

Inchi Code

1S/C12H9BrO/c13-11-6-10(7-12(14)8-11)9-4-2-1-3-5-9/h1-8,14H

InChI key

JCQWHMKUEMXGQC-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-5121

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-5121


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library