E-504
Estrogens CRM in Serum solution
50 pg/mL (Sigmatrix Ultra), certified reference material, vial of 1 mL
About This Item
Recommended Products
grade
certified reference material
feature
Snap-N-Spike®/Snap-N-Shoot®
composition
17ß-Estradiol, 500 pg/mL (each analyte)
Estrone, 500 pg/mL (each analyte)
packaging
vial of 1 mL
manufacturer/tradename
Cerilliant®
concentration
50 pg/mL (Sigmatrix Ultra)
application(s)
clinical testing
format
multi-component solution
storage temp.
−20°C
General description
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
E-504-1ML:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service