Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

999992C

Avanti

16:0 Diether PC

Avanti Research - A Croda Brand 999992C

Synonym(s):

1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H84NO6P
CAS Number:
Molecular Weight:
706.07
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (999992C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999992C

concentration

10 mg/mL (999992C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCCCCCC)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C40H84NO6P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-35-44-38-40(39-47-48(42,43)46-37-34-41(3,4)5)45-36-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3

InChI key

MWTIGLPPQBNUFP-UHFFFAOYSA-N

General description

16:0 Diether PC (phosphocholine) is a zwitterionic lipid having two hexadecane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.

Application

16:0 Diether PC or 1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine might be used:
  • to examine the miscibility and phase behavior of the cationic 1,2-dipalmitoyl-sn-glycero-3-ethylphosphocholine (EDPPC)
  • as a component of unilamellar vesicles to study glycolipid transfer protein (GLTP)-mediated transfer of anthrylvinyl-galactosylceramide (AV-GalCer) from different ether phospholipid matrices
  • to compose large phospholipid bicelles to determine the three-dimensional backbone structure of the transmembrane domain of Vpu from human immunodeficiency virus (HIV)-1 by solid-state nuclear magnetic resonance (NMR) spectroscopy
  • as a non-ladderane phosphocholine (PC) lipid reference sample in reversed-phase liquid chromatography (LC) method to separate PC lipids

Biochem/physiol Actions

Ether linked phospholipids serve as precursors for important signaling molecules. Ether glycerolipids act as membrane molecules and cellular mediators. 16:0 Diether PC (phosphocholine) interdigitates even in the absence of chemical inducers or pressure.

Packaging

5 mL Clear Glass Sealed Ampule (999992C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

999992C-10MG:
999992C-VAR:
999992C-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ether-linked lipids and their bioactive species
Biochemistry of Lipids, Lipoproteins and Membranes, 9(2), 245-276 (2008)
The interdigitated gel phase in mixtures of cationic and zwitterionic phospholipids
Smith EA and Dea PH
Biophysical Chemistry, 196(2), 86-91 (2015)
Analysis of intact ladderane phospholipids, originating from viable anammox bacteria, using RP-LC-ESI-MS
Lanekoff I and Karlsson R
Analytical and Bioanalytical Chemistry, 397(8), 3543-3551 (2010)
Three-dimensional structure of the transmembrane domain of Vpu from HIV-1 in aligned phospholipid bicelles
Park SH, et al.
Biophysical Journal, 91(8), 3032-3042 (2006)
Molecular features of phospholipids that affect glycolipid transfer protein-mediated galactosylceramide transfer between vesicles
Nylund M, et al.
Biochimica et Biophysica Acta - Biomembranes, 1758(6), 807-812 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service