Skip to Content
Merck
All Photos(1)

Documents

Safety Information

860960M

Avanti

Trifunctional Sphingosine

Avanti Research - A Croda Brand

Synonym(s):

N-Diethylaminocoumarinacarbonyl photoclick sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H48N4O6
CAS Number:
Molecular Weight:
608.77
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

methylene chloride solution

packaging

package of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

250 μg/mL in methylene chloride

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H](NC(OCC(C(C=CC(N(CC)CC)=C1)=C1O2)=CC2=O)=O)[C@H](O)/C=C/CCCCCCCCC3(N=N3)CCCC#C

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

860960M-250UG:
860960M-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service