Skip to Content
Merck
All Photos(1)

Documents

Safety Information

860852P

Avanti

CER9 (t18:0/26:0/18:1(d9))

Avanti Research - A Croda Brand 860852P, powder

Synonym(s):

N-[26-oleoyloxy(d9) hexacosanoyl]-D-ribo-phytosphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H112D9NO6
CAS Number:
Molecular Weight:
985.68
UNSPSC Code:
12352100
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860852P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860852P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(N[C@H]([C@H](O)[C@H](O)CCCCCCCCCCCCCC)CO)CCCCCCCCCCCCCCCCCCCCCCCCCOC(CCCCCCC/C=C\CCCCC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])=O

Packaging

5 mL Amber Glass Screw Cap Vial (860852P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

860852P-1MG:
860852P-BULK:
860852P-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service