Skip to Content
Merck
All Photos(2)

Documents

Safety Information

860453C

Avanti

16:0-d31-18:1 PA

1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

110920

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H39O8PNaD31
CAS Number:
Molecular Weight:
728.10
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860453C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860453C

concentration

10 mg/mL (860453C-10mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C37H71O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35H,3-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b18-17-;/t35-;/m1./s1/i2D3,4D2,6D2,8D2,10D2,12D2,14D2,16D2,19D2,21D2,23D2,25D2,27D2,29D2,31D2;

InChI key

PPYXMAJBOKWTQX-VYSSPCEXSA-M

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31-18:1 PA has been used as an internal standard to quantify fatty acid methyl esters (FAMES) using electrospray ionization tandem mass spectrometry (ESI-MS/MS)-based lipidomic analysis.

Packaging

5 mL Clear Glass Sealed Ampule (860453C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

860453C-VAR:
860453C-BULK:
860453C-10MG:4548174018545


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delineating the rules for structural adaptation of membrane-associated proteins to evolutionary changes in membrane lipidome
Makarova M, et al.
Current Biology (2020)
Jordon M Inloes et al.
Biochemistry, 57(39), 5759-5767 (2018-09-18)
Deleterious mutations in the serine hydrolase DDHD domain containing 1 (DDHD1) cause the SPG28 subtype of the neurological disease hereditary spastic paraplegia (HSP), which is characterized by axonal neuropathy and gait impairments. DDHD1 has been shown to display PLA1-type phospholipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service