Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

810212C

Avanti

C6-NBD Sphinganine

Avanti Research - A Croda Brand 810212C

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
¥19,650

¥19,650


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
¥19,650

About This Item

Empirical Formula (Hill Notation):
C30H51N5O6
CAS Number:
Molecular Weight:
577.76
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

¥19,650


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810212C-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810212C

concentration

0.05 mg/mL (810212C-50ug)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1c2n[o]nc2c(cc1)NCCCCCC(=O)N[C@H]([C@H](O)CCCCCCCCCCCCCCC)CO

InChI

1S/C30H51N5O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-27(37)25(23-36)32-28(38)19-16-14-17-22-31-24-20-21-26(35(39)40)30-29(24)33-41-34-30/h20-21,25,27,31,36-37H,2-19,22-23H2,1H3,(H,32,38)/t25-,27+/m0/s1

InChI key

GEZLBJRDZRUTOE-AHKZPQOWSA-N

General description

C6-Sphinganine, also called as C6-dihydrosphingosine, is a common sphingoid base in phospholipids with C6 acyl chain.[1] C6-NBD Sphinganine is an omega fatty acid labeled fluorescent sphingolipid.

Packaging

5 mL Amber Glass Screw Cap Vial (810212C-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

810212C-50UG:
810212C-VAR:
810212C-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Thevissen et al.
Bioorganic & medicinal chemistry letters, 18(13), 3728-3730 (2008-06-06)
The minimal fungicidal concentration (MFC) of dihydrosphingosine (DHS), phytosphingosine (PHS), and five short-chain DHS derivatives was determined for Candida albicans and Candida glabrata. In this respect, a C15- and a C17-homologue of DHS showed a 2- to 10-fold decreased MFC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service