700339P
Avanti
Glyco-ω-muricholanoic acid
Avanti Research™ - A Croda Brand
Synonym(s):
5ß-Cholanic acid-3α,6α,7ß-triol N-(carboxymethyl)-amide
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
package of 1 × 1 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
bile acids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[H][C@@]12[C@]([C@](CC[C@@H](O)C3)(C)[C@]3([H])[C@@H](O)[C@@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(NCC(O)=O)=O
InChI
1S/C26H43NO6/c1-14(4-7-20(29)27-13-21(30)31)16-5-6-17-22-18(9-11-25(16,17)2)26(3)10-8-15(28)12-19(26)23(32)24(22)33/h14-19,22-24,28,32-33H,4-13H2,1-3H3,(H,27,29)(H,30,31)/t14-,15-,16-,17+,18+,19+,22+,23-,24-,25-,26-/m1/s1
InChI key
ZQYUKJFJPJDMMR-CINIKCAZSA-N
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
700339P-1MG:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service