Skip to Content
Merck
All Photos(1)

Documents

Safety Information

330824L

Avanti

SM Internal Standard Mixture - UltimateSPLASH

Avanti Research - A Croda Brand

Synonym(s):

SM Internal Standard Mixture - UltimateSPLASH for Lipidomic Analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

solution

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (330824L-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 860741L16:1 SM (d18:1/16:1)-d9, Avanti Research - A Croda Brand 75μg/mLSDS

  • 860740L18:1 SM (d18:1/18:1)-d9, Avanti Research - A Croda Brand 860740L-1MG
    860746L-1MG 50μg/mL
    SDS

  • 860742L20:1 SM (d18:1/20:1)-d9, Avanti Research - A Croda Brand 25μg/mLSDS

  • 860743L22:1 SM (d18:1/22:1)-d9, Avanti Research - A Croda Brand 50μg/mLSDS

  • 860744L24:1 SM (d18:1/24:1)-d9, Avanti Research - A Croda Brand 75μg/mLSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

330824L-1EA:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service