Skip to Content
Merck
All Photos(1)

Key Documents

ATE514313369

AstaTech, Inc.

METHYL 3,5-DICHLORO-4-FORMYLBENZOATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.05
MDL number:

Iupac Name

methyl 3,5-dichloro-4-formylbenzoate

Inchi Code

1S/C9H6Cl2O3/c1-14-9(13)5-2-7(10)6(4-12)8(11)3-5/h2-4H,1H3

InChI key

PZMMESDUAFUZSK-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=77846

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=CL9613


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library