Skip to Content
Merck
All Photos(1)

Key Documents

ATE429352127

AstaTech, Inc.

TERT-BUTYL 3-(4-BROMOPHENYL)PYRROLIDINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
326.23
MDL number:

Iupac Name

tert-butyl 3-(4-bromophenyl)-1-pyrrolidinecarboxylate

Inchi Code

1S/C15H20BrNO2/c1-15(2,3)19-14(18)17-9-8-12(10-17)11-4-6-13(16)7-5-11/h4-7,12H,8-10H2,1-3H3

InChI key

NFTKVQYDQFSNHQ-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=48936

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=88148


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library