AMBH303C5200
Ambeed, Inc.
3-Chloro-4-nitrobenzene-1-sulfonyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-chloro-4-nitrobenzenesulfonyl chloride
Inchi Code
1S/C6H3Cl2NO4S/c7-5-3-4(14(8,12)13)1-2-6(5)9(10)11/h1-3H
InChI key
BYKPVHSGMXQEEY-UHFFFAOYSA-N
storage temp.
Inert atmosphere,2-8C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Solid or Semi-Solid or Liquid or Lump
Product Link
https://www.ambeed.com/products/64835-30-9.html
Signal Word
Danger
Hazard Statements
Precautionary statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.