Key Documents
Z743269
Julabo® tube clamp
size 3, for 1/2 inch i.d. reinforced tubing
Select a Size
2 EA
¥5,712
¥5,712
Estimated to ship onMay 31, 2025
Select a Size
Change View
2 EA
¥5,712
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
¥5,712
Estimated to ship onMay 31, 2025
packaging
pack of 2 ea
manufacturer/tradename
Julabo 8970482
General description
Tube clamps for ensuring secure fixing of tubing from the application to the temperature control instrument.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)