Z740681
Aldrich® Essentials filter flask, with side arm
capacity 250 mL
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass flask
flat bottom flask
Joint
joint stopper6
packaging
pkg of 6 ea
capacity
250 mL
Related Categories
General description
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC